Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vsx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131462.2 | NP_571537.2 | 2 - 3 | 579 | 106 | |
XM_005158805.4 | XP_005158862.1 | 2 - 3 | 824 | 106 | ||
GenBank mRNA | BC055588.1 | - | 2 - 3 | 579 | 106 | |
BC067664.1 | - | 2 - 3 | 541 | 106 | ||
BC164202.1 | - | 2 - 3 | 541 | 106 | ||
U62898.1 | - | 2 - 3 | 573 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |