Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cldni | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131768.2 | NP_571843.1 | 2 - 3 | 245 | 102 | |
GenBank mRNA | AF359428.1 | - | 2 - 3 | 231 | 102 | |
BC046020.1 | - | 2 - 3 | 242 | 102 | ||
BC065448.1 | - | 2 - 3 | 230 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |