Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: col1a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199214.1 | NP_954684.1 | 34 - 35 | 2350 | 75 | |
GenBank mRNA | AY380817.1 | - | 34 - 35 | 2350 | 75 | |
BC063249.1 | - | 34 - 35 | 2445 | 75 | ||
BC161663.1 | - | 34 - 35 | 2462 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |