Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cd63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199543.1 | NP_955837.1 | 6 - 7 | 632 | 71 | |
GenBank mRNA | BC056698.1 | - | 6 - 7 | 632 | 71 | |
BC064300.1 | - | 6 - 7 | 670 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |