Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smarce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201298.1 | NP_958455.2 | 7 - 8 | 612 | 74 | |
XM_005163989.4 | XP_005164046.1 | 7 - 8 | 708 | 74 | ||
XM_005163990.4 | XP_005164047.1 | 7 - 8 | 696 | 74 | ||
XM_009299517.3 | XP_009297792.1 | 7 - 8 | 702 | 74 | ||
XM_021469404.1 | XP_021325079.1 | 6 - 7 | 603 | 74 | ||
XM_021469407.1 | XP_021325082.1 | 6 - 7 | 591 | 74 | ||
GenBank mRNA | BC044363.1 | - | 7 - 8 | 612 | 74 | |
BC066435.1 | - | 6 - 7 | 588 | 74 | ||
BC164725.1 | - | 5 - 6 | 576 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |