Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kpna7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213070.1 | NP_998235.1 | 5 - 6 | 780 | 58 | |
GenBank mRNA | BC044523.1 | - | 5 - 6 | 780 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |