Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aven | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045292.2 | NP_001038757.2 | 6 | 2300 | 120 | |
GenBank mRNA | BC116469.1 | - | 6 | 2251 | 120 |
Gene Symbol: ryr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009294773.3 | XP_009293048.1 | 103 | 15158 | 120 | |
XM_009294774.3 | XP_009293049.1 | 102 | 15140 | 120 | ||
XM_009294775.3 | XP_009293050.1 | 102 | 15137 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Both primers and probe map within a single exon |