Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klhl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC121773.1 | - | 2 | 752 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Both primers and probe map within a single exon |