Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crygm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007785.1 | NP_001007786.1 | 3 | 545 | 113 | |
GenBank mRNA | AY738749.1 | - | 3 | 545 | 113 | |
BC133864.1 | - | 3 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |