Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sat1a.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002169.2 | NP_001002169.1 | 6 | 1271 | 89 | |
GenBank mRNA | BC072720.1 | - | 6 | 1254 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |