Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: foxj1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008648.1 | NP_001008648.1 | 2 | 449 | 142 | |
XM_009306612.2 | XP_009304887.1 | 1 | 699 | 142 | ||
GenBank mRNA | BC086726.1 | - | 2 | 449 | 142 | |
BC165812.1 | - | 1 | 201 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |