Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004582.1 | NP_001004582.1 | 3 - 4 | 243 | 100 | |
GenBank mRNA | BC081638.1 | - | 3 - 4 | 243 | 100 | |
BC153573.1 | - | 3 - 4 | 243 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |