Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adprhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006033.2 | NP_001006033.1 | 3 - 4 | 549 | 57 | |
XM_005167996.4 | XP_005168053.1 | 3 - 4 | 546 | 57 | ||
XM_021473006.1 | XP_021328681.1 | 3 - 4 | 640 | 57 | ||
GenBank mRNA | BC083385.1 | - | 3 - 4 | 549 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |