Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: enpp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013527.1 | NP_001013545.1 | 3 - 4 | 656 | 95 | |
GenBank mRNA | BC090903.1 | - | 3 - 4 | 656 | 95 | |
BC165573.1 | - | 3 - 4 | 557 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |