Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nsdhl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017674.1 | NP_001017674.1 | 7 - 8 | 794 | 68 | |
GenBank mRNA | BC093332.1 | - | 7 - 8 | 794 | 68 | |
BC164386.1 | - | 6 - 7 | 736 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |