Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrrc39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017760.1 | NP_001017760.1 | 5 - 6 | 785 | 146 | |
GenBank mRNA | BC093194.1 | - | 5 - 6 | 785 | 146 | |
BC153572.1 | - | 5 - 6 | 582 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |