Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: qdprb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020698.1 | NP_001018534.1 | 6 - 7 | 651 | 72 | |
GenBank mRNA | BC081416.1 | - | 6 - 7 | 650 | 72 | |
BC095805.1 | - | 6 - 7 | 651 | 72 | ||
BC165162.1 | - | 6 - 7 | 652 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |