Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrpl28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024399.1 | NP_001019570.1 | 2 - 3 | 402 | 75 | |
GenBank mRNA | BC095131.1 | - | 2 - 3 | 402 | 75 | |
BC154265.1 | - | 2 - 3 | 353 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |