Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nr3c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100403.1 | NP_001093873.1 | 3 - 4 | 2217 | 79 | |
GenBank mRNA | EF567113.1 | - | 3 - 4 | 2217 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |