Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrpl23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002052.2 | NP_001002052.1 | 2 - 3 | 202 | 147 | |
GenBank mRNA | BC071325.1 | - | 2 - 3 | 203 | 147 | |
BC153659.1 | - | 2 - 3 | 234 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |