Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mthfd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002181.1 | NP_001002181.1 | 6 - 7 | 906 | 89 | |
GenBank mRNA | BC074043.1 | - | 6 - 7 | 906 | 89 | |
DQ100356.1 | - | 6 - 7 | 841 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |