Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btbd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002198.1 | NP_001002198.1 | 7 - 8 | 1441 | 57 | |
XM_021480593.1 | XP_021336268.1 | 7 - 8 | 1508 | 57 | ||
GenBank mRNA | BC074031.1 | - | 7 - 8 | 1441 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |