Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gadd45ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002216.1 | NP_001002216.1 | 2 - 3 | 262 | 62 | |
GenBank mRNA | BC074023.1 | - | 2 - 3 | 234 | 62 | |
BC075932.1 | - | 2 - 3 | 262 | 62 | ||
BC152637.1 | - | 2 - 3 | 254 | 62 | ||
BC164910.1 | - | 2 - 3 | 181 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |