Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: galm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002373.1 | NP_001002373.1 | 6 - 7 | 1029 | 54 | |
GenBank mRNA | BC076039.1 | - | 6 - 7 | 1029 | 54 | |
BC153585.1 | - | 6 - 7 | 1022 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |