Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myoz1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002447.1 | NP_001002447.1 | 4 - 5 | 399 | 91 | |
XM_005156283.4 | XP_005156340.1 | 4 - 5 | 517 | 91 | ||
XM_005156285.3 | XP_005156342.1 | 4 - 5 | 525 | 91 | ||
XM_017358446.2 | XP_017213935.1 | 4 - 5 | 441 | 91 | ||
GenBank mRNA | BC076024.1 | - | 4 - 5 | 399 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |