Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrps7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033596.1 | NP_001028768.1 | 1 - 2 | 76 | 147 | |
GenBank mRNA | BC100013.1 | - | 1 - 2 | 76 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |