Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: etnk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034175.1 | NP_001029347.1 | 5 - 6 | 966 | 154 | |
GenBank mRNA | BC096922.1 | - | 5 - 6 | 966 | 154 | |
BC164400.1 | - | 5 - 6 | 807 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |