Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fktn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042694.2 | NP_001036159.2 | 4 - 5 | 536 | 90 | |
XM_021475968.1 | XP_021331643.1 | 4 - 5 | 704 | 90 | ||
GenBank mRNA | BC134110.1 | - | 4 - 5 | 536 | 90 | |
DQ826745.1 | - | 3 - 4 | 424 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |