Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nab1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045086.1 | NP_001038551.1 | 3 - 4 | 967 | 66 | |
XM_005161686.4 | XP_005161743.1 | 4 - 5 | 1234 | 66 | ||
XM_009296205.3 | XP_009294480.1 | 4 - 5 | 1237 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |