Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: amn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045454.1 | NP_001038919.1 | 6 - 7 | 817 | 90 | |
GenBank mRNA | BC122149.1 | - | 6 - 7 | 817 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |