Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076619.1 | NP_001070087.1 | 6 - 7 | 1473 | 73 | |
XM_017356912.2 | XP_017212401.1 | 6 - 7 | 1589 | 73 | ||
GenBank mRNA | BC124441.1 | - | 6 - 7 | 1473 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |