Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: enosf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076742.2 | NP_001070210.2 | 8 - 9 | 691 | 86 | |
GenBank mRNA | BC124261.1 | - | 8 - 9 | 691 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |