Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: palm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080085.1 | NP_001073554.1 | 6 - 7 | 771 | 124 | |
GenBank mRNA | BC128825.1 | - | 6 - 7 | 771 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |