Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ribc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080663.2 | NP_001074132.2 | 5 - 6 | 1168 | 94 | |
GenBank mRNA | BC129188.1 | - | 5 - 6 | 1173 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |