Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ddx3xa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126423.1 | NP_001119895.1 | 15 - 16 | 2053 | 108 | |
XM_005167718.4 | XP_005167775.1 | 16 - 17 | 2157 | 108 | ||
XM_005167720.4 | XP_005167777.1 | 14 - 15 | 2049 | 108 | ||
XM_009304751.3 | XP_009303026.1 | 15 - 16 | 2094 | 108 | ||
XM_017357705.2 | XP_017213194.1 | 16 - 17 | 2154 | 108 | ||
XM_017357706.2 | XP_017213195.1 | 16 - 17 | 2154 | 108 | ||
XM_021478726.1 | XP_021334401.1 | 18 - 19 | 2494 | 108 | ||
XM_021478727.1 | XP_021334402.1 | 18 - 19 | 2491 | 108 | ||
XM_021478728.1 | XP_021334403.1 | 18 - 19 | 2491 | 108 | ||
XM_021478729.1 | XP_021334404.1 | 17 - 18 | 2446 | 108 | ||
XM_021478730.1 | XP_021334405.1 | 17 - 18 | 2434 | 108 | ||
XM_021478731.1 | XP_021334406.1 | 17 - 18 | 2410 | 108 | ||
XM_021478733.1 | XP_021334408.1 | 19 - 20 | 2407 | 108 | ||
XM_021478734.1 | XP_021334409.1 | 18 - 19 | 2347 | 108 | ||
XM_021478735.1 | XP_021334410.1 | 18 - 19 | 2323 | 108 | ||
XM_021478736.1 | XP_021334411.1 | 15 - 16 | 2109 | 108 | ||
GenBank mRNA | BC133162.1 | - | 15 - 16 | 2053 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |