Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arid3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083002.1 | NP_001076471.1 | 6 - 7 | 1348 | 121 | |
GenBank mRNA | BC131867.1 | - | 6 - 7 | 1348 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |