Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: taar14g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083055.1 | NP_001076524.1 | 1 - 2 | 764 | 75 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Recommended Assay Danio rerio Probe spans exons |