Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhcga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089577.1 | NP_001083046.1 | 4 - 5 | 738 | 62 | |
GenBank mRNA | AB286867.1 | - | 4 - 5 | 763 | 62 | |
BC139524.1 | - | 4 - 5 | 738 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |