Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: and2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111190.1 | NP_001104660.1 | 3 - 4 | 186 | 69 | |
GenBank mRNA | BC155829.1 | - | 3 - 4 | 186 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |