Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pi4kab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113342.1 | NP_001106813.1 | 10 - 11 | 1170 | 83 | |
GenBank mRNA | BC155592.1 | - | 10 - 11 | 1170 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |