Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: il23r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113506.1 | NP_001106978.1 | 6 - 7 | 992 | 93 | |
XM_021476745.1 | XP_021332420.1 | 9 - 10 | 1874 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons |