Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113611.1 | NP_001107083.1 | 3 - 4 | 403 | 66 | |
GenBank mRNA | BC154468.1 | - | 3 - 4 | 403 | 66 | |
BC171683.1 | - | 3 - 4 | 398 | 66 | ||
BC171685.1 | - | 3 - 4 | 398 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |