Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phactr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114411.1 | NP_001107883.1 | 10 - 11 | 1526 | 76 | |
GenBank mRNA | BC154544.1 | - | 10 - 11 | 1526 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |