Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cldn12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131773.2 | NP_571848.1 | 2 - 3 | 464 | 71 | |
GenBank mRNA | AF359431.1 | - | 2 - 3 | 384 | 71 | |
AF359433.1 | - | 2 - 3 | 425 | 71 | ||
BC075744.1 | - | 2 - 3 | 428 | 71 | ||
EH545623.1 | - | 2 - 3 | 477 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |