Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbx18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153665.1 | NP_705951.1 | 2 - 3 | 617 | 85 | |
GenBank mRNA | AF448503.3 | - | 2 - 3 | 605 | 85 | |
BC154826.1 | - | 2 - 3 | 601 | 85 | ||
BC163460.1 | - | 2 - 3 | 562 | 85 | ||
BC163482.1 | - | 2 - 3 | 562 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |