Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fgf8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182856.1 | NP_878276.1 | 4 - 5 | 851 | 72 | |
GenBank mRNA | AB100171.1 | - | 4 - 5 | 851 | 72 | |
BC163345.1 | - | 4 - 5 | 612 | 72 | ||
BC163362.1 | - | 4 - 5 | 612 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |