Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atad5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082829.2 | NP_001076298.2 | 13 - 14 | 3227 | 84 | |
XM_005166177.4 | XP_005166234.1 | 13 - 14 | 3283 | 84 | ||
GenBank mRNA | BC057535.1 | - | 13 - 14 | 3227 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |