Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199647.2 | NP_955941.2 | 4 - 5 | 732 | 91 | |
GenBank mRNA | BC059596.1 | - | 4 - 5 | 669 | 91 | |
EH546236.1 | - | 4 - 5 | 771 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |