Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hirip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199823.1 | NP_956117.1 | 7 - 8 | 1377 | 60 | |
GenBank mRNA | BC055674.1 | - | 7 - 8 | 1377 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |