Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199824.1 | NP_956118.1 | 1 - 2 | 307 | 72 | |
GenBank mRNA | BC052766.1 | - | 1 - 2 | 307 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |